Titriplex® III (ethylenedinitrilotetraacetic acid disodium salt dihydrate), EMPROVE® EXPERT ACS, JP, USP, BP, Ph. Eur. for biopharmaceutical production, SAFC®
Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Ethylenediaminetetraacetic acid, disodium salt, dihydrate, Ethylenediaminetetraacetic acid disodium salt dihydrate, Kestranal® 2S dihydrate, Disodium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid disodium salt dihydrate, Ethylenediaminetetraacetic acid disodium dihydrate, Titriplex III, Ethylenediaminetetra-acetic acid disodium salt dihydrate, Kestranal® 2S 2 H2O, Sequestrene Na₂, Edetate disodium dihydrate, Edetic acid disodium salt dihydrate
1.37004.1000 1.37004.9012 1.37004.9025
1.37004.1000EA 2800 DKK
1.37004.1000 1.37004.9012 1.37004.9025
Titriplex® III (ethylenedinitrilotetraacetic acid disodium salt dihydrate), EMPROVE® EXPERT ACS, JP, USP, BP, Ph. Eur. for biopharmaceutical production, SAFC®
EDTA dinatriumsalt dihydrat
Formel: C₁₀H₁₄N₂Na₂O₈·2H₂O
Molekylvægt: 372.24 g/mol
Smeltepunkt: 255
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00150037
CAS-nummer: 6381-92-6
EINECS: 205-358-3
Merck Index: 13,03543
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM