tri-Natriumcitrat, anhydrous, EMPROVE® ESSENTIAL FCC, USP, E331 suitable for use as excipient, SAFC®
Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Sodium Citrate, Sodium citrate tribasic, Citric acid trisodium salt, Sodium 2-hydroxypropane-1,2,3-tricarboxylate
1.11037.9026 1.11037.1000
1.11037.9026EA 14560 DKK
1.11037.9026 1.11037.1000
tri-Natriumcitrat, anhydrous, EMPROVE® ESSENTIAL FCC, USP, E331 suitable for use as excipient, SAFC®
tri-Natriumcitrat
Formel: C₆H₅Na₃O₇
Molekylvægt: 258.07 g/mol
Smeltepunkt: 150
Massefylde: 1.815 g/cm³ (20 °C)
Flammepunkt: 290
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00012462
CAS-nummer: 68-04-2
EINECS: 200-675-3
Merck Index: 12,08746