Oxalsyre dihydrat
Leverandør: Roth Carl
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
Ethanedioic acid dihydrate
8879.1
8879.2
ROTH8879.1EA
0
DKK
ROTH8879.1
ROTH8879.2
Oxalsyre dihydrat
Oxalsyre dihydrat
Formel:
HO₂CCO₂H·2H₂O Molekylvægt: 126.07 g/mol Kogepunkt: 149…160 °C (1013 hPa) Smeltepunkt: 101 Massefylde: 1.65 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00149102 CAS-nummer: 6153-56-6 EINECS: 205-634-3 UN nummer: 3261 ADR: 8 III REACH: 01-2119534576-33 Merck Index: 13,06980 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...