Kieselgel, TECHNICAL drying agent, with moisture indicator cobalt (II) chloride (blue gel)
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
92120.0120EA 0 DKK
92120.0120 92120.9012
Kieselgel, TECHNICAL drying agent, with moisture indicator cobalt (II) chloride (blue gel)
Silica gel
Formel: SiO₂
Molekylvægt: 60.08 g/mol
Kogepunkt: 2230
Smeltepunkt: 1710
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011232
CAS-nummer: 63231-67-4
Bestil nu
Specifikationer Testresultater
Appearance Blue granules
Loss on drying (145°C) Max. 2 %
Absorption capacity (100% RH) Min. 30 %