Guanidiniumchlorid

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Guanidinhydrochlorid

39558.01 39558.02
SERA39558.01EA 0 DKK
SERA39558.01 SERA39558.02
Guanidiniumchlorid
Guanidiniumchlorid
Formel: NH₂C(=NH)NH₂·HCl
Molekylvægt: 95.53 g/mol
Smeltepunkt: 185
Massefylde: 1.354 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013026
CAS-nummer: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR