Guanidiniumcarbonat ≥98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Guanidine carbonate salt, Diguanidinium carbonate, Guanidine carbonate

A12853.0B A12853.0I A12853.30
A12853.0BEA 733 DKK
A12853.0B A12853.0I A12853.30
Guanidiniumcarbonat ≥98%
Guanidiniumcarbonat

Purity: 98% CAS No.: 593-85-1 Molecular Formula: C3H12N6O3 Formula Weight: 180.17 Melting Point: ca 198° dec. Beilstein Reference: 3628359

Formel: C₃H₁₂N₆O₃
Molekylvægt: 180.17 g/mol
Smeltepunkt: ∼ 198 °C (dec.)
Massefylde: 1.251 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00013029
CAS-nummer: 593-85-1
EINECS: 209-813-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR