Dimethylammoniumchlorid for synthesis, Sigma-Aldrich®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Dimethylamine hydrochloride
M-Clarity™ quality level = MQ200
8.03052.0005 8.03052.0250 8.03052.1000 8.03052.9025
8.03052.0005EA 200 DKK
8.03052.0005 8.03052.0250 8.03052.1000 8.03052.9025
Dimethylammoniumchlorid for synthesis, Sigma-Aldrich®
Dimethylammoniumchlorid
Formel: C₂H₈ClN
Molekylvægt: 81.54 g/mol
Smeltepunkt: 170…173 °C
MDL Number: MFCD00012477
CAS-nummer: 506-59-2
EINECS: 208-046-5
Merck Index: 13,03255
Bestil nu
Specifikationer Testresultater
Solubility 3000 g/l (20 °C)
Melting Point 160 °C
Molar Mass 81.54 g/mol
Bulk Density 500 kg/m3
Vapor Pressure  - 0.1 hPa (25 °C)
pH 5 (100 g/l, H2O, 20 °C)