L(+)-Cystein, HCl monohydrat
Leverandør: Roth Carl
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: (R)-(+)-Cysteine hydrochloride monohydrate, (R)-2-Amino-3-mercaptopropanoic acid hydrochloride hydrate, L-Cysteine HCI Monohydrate, H-Cys-OH.HCl.H2O, Cysteine Hydrochloride, Monohydrate, Cysteine Hydrochloride monohydrate
T203.1 T203.2 T203.3
ROTHT203.1EA 237.93 DKK
ROTHT203.1 ROTHT203.2 ROTHT203.3
L(+)-Cystein, HCl monohydrat
L(+)-Cystein, HCl monohydrat
Formel: HSCH₂CH(NH₂)COOH·HCl·H₂O
Molekylvægt: 175.64 g/mol
Smeltepunkt: 168…170 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00065606
CAS-nummer: 7048-04-6
EINECS: 200-157-7
Merck Index: 13,02810
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18703c57a6b5f299a5f38f5d92b333d3d65de497&vid=10395059845247050509d84df67aefcb3c98806c&prid=web&clid=SAPDAM