Natriumhydroxid 30% (w/w) in aqueous solution
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Caustic soda
87864.AE 87864.AP
87864.AEEA 385 DKK
87864.AE 87864.AP
Natriumhydroxid 30% (w/w) in aqueous solution
Sodium hydroxide
CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Physical Form: Liquid Appearance: Colourless UN No.: 1824 MDL No.: MFCD00003548
Formel: NaOH
Molekylvægt: 40 g/mol
Kogepunkt: 119…135 °C (1013 hPa)
Smeltepunkt: 9…10 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
UN nummer: 1824
ADR: 8 II
Merck Index: 14,08627