Ethidiumbromid 10 mg/ml aqueous solution, solution_dark red Molecular_Biology
Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: 3,8-Diamino-5-ethyl-6-phenylphenantridiniumbromid
ETBC1002 11ETBC1001 11ETBC1002
QBIOETBC1002EA 0 DKK
QBIOETBC1002 ICNA11ETBC1001 ICNA11ETBC1002
Ethidiumbromid 10 mg/ml aqueous solution, solution_dark red Molecular_Biology
Ethidiumbromid
A 10 mg/mL Ready-To-Use solution of EtBr in specially filtered, deionized water.
  • Grade : Molecular Biology
  • Concentration : (10 mg/ml)
  • Suitability : Suitable for gel electrophoresis
  • Key Applications : Nucleic acid stain for PAGE or agarose gel electrophoresis; Used in fluorimetric assays for nucleic acids
  • Application Areas : Nucleic Acid Electrophoresis; Blotting & Detection; Nucleic Acid Gel Electrophoresis & Blotting
  • Product Type : Biochemicals
  • Analyte(s) Detected : dsDNA; dsRNA
  • Biochemical Category : Labels, Dyes & Stains
  • Chemical Class : Labels, Dyes & Stains
  • Vapor Pressure : 1.2 X 10-12 mm Hg at 25 °C
  • UV/Visible Absorbance : 254 nm
  • Presentation : Dark Red Solution
  • Detection Method : UV gel box (254 nm)
  • Solubility : Soluble in water.
  • Recommended Replacement For : Ethidium Bromide Powder
  • Certifications : DNase, RNase Protease Free
  • Detection/Incubation Time : 15-30 min with 0.5 mg/mL solution
Storage: protect from light.
Formel: C₂₁H₂₀BrN₃
Molekylvægt: 394.31 g/mol
Kogepunkt: 72
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00011724
CAS-nummer: 1239-45-8
EINECS: 214-984-6
UN nummer: 2810
Merck Index: 13,04751
Bestil nu
Specifikationer Testresultater
Identity Test Passes