Salpetersyre 65 - 70% 99,9999 Metals basis
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Nitric acid solution
10984.AC 10984.AK 10984.AP
10984.ACEA 286 DKK
10984.AC 10984.AK 10984.AP
Salpetersyre 65 - 70% 99,9999 Metals basis
Salpetersyre
Purity: 99.9999% CAS No.: 7697-37-2 Molecular Formula: HNO3 Formula Weight: 63.01 Physical Form: Liquid Density: 1.41 Appearance: Colourless to pale yellow Odour: Pungent Boiling Point: 121° Merck Reference: 14,6577 UN No.: 2031 MDL No.: MFCD00011349
Formel: HNO₃
Molekylvægt: 63.01 g/mol
Kogepunkt: 121 °C
Smeltepunkt: –42 °C
Massefylde: 1.41
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011349
CAS-nummer: 7697-37-2
EINECS: 231-714-2
UN nummer: 2031
ADR: 8 II
Merck Index: 14,6577