Cerium (IV) sulfat 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur., Reag. USP volumetric solution
Leverandør: VWR Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: Cerium disulphate, Ceric sulphate, Cerium(IV) sulphate
190474MEA 1070 DKK
190474M 190475N
Cerium (IV) sulfat 0.1 mol/l (0.1 N) in aqueous solution, AVS TITRINORM Reag. Ph. Eur., Reag. USP volumetric solution
Cerium (IV) sulfat
Formel: Ce(SO₄)₂
Molekylvægt: 332.24 g/mol
Smeltepunkt: 195
MDL Number: MFCD00148852
CAS-nummer: 13590-82-4
EINECS: 237-029-5
UN nummer: 3264
ADR: 8 III
Merck Index: 12,02036
Bestil nu
Specifikationer Testresultater
Titer (20°C; real value 0.3 % accuracy) 0,0995-0,1005 mol/l
NIST traceable Confirmed
Conforms to Reag. Ph.Eur. Passes test
Conforms to Reag. USP Passes test