Saltsyre 37% ACS, Fluka™
Leverandør: Honeywell Chemicals
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
320331-500ML 320331-6X500ML
320331-500MLEA 827 DKK
320331-500ML 320331-6X500ML
Saltsyre 37% ACS, Fluka™
Saltsyre
Formel: HCl
Kogepunkt: 42 °C (1,013 hPa)
Smeltepunkt: –35 °C
Massefylde: 1,190 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
UN nummer: 1789
ADR: 8,II
Bestil nu
Specifikationer Testresultater
Assay 36.5 - 38.0 %
Residue on ignition (SO₄) Max. 5 ppm
Arsenic (As) Max. 0.01 ppm
Vapor Pressure 3.23 psi ( 21.1 °C)
Iron (Fe) Max. 0.2 ppm
Vapor Density 1.3 (vs air)
Heavy metals (ICP) ACS Max. 1 ppm
Ammonium (NH₄) Max. 1 ppm
Bromide (Br) Max. 50 ppm
Free chlorine (Cl) Max. 1 ppm
Sulfate (SO₄) Max. 1 ppm
Sulfite (SO₃) Max. 1 ppm
APHA Max. 10