Saltsyre fuming 37%, EMPROVE® EXPERT Ph. Eur., ACS, JP, NF, BP for biopharmaceutical production, SAFC®
Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Hydrochloric acid solution, Muriatic acid
1.37007.2500 1.37007.9025 1.37007.1000
1.37007.2500EA 1480 DKK
1.37007.2500 1.37007.9025 1.37007.1000
Saltsyre fuming 37%, EMPROVE® EXPERT Ph. Eur., ACS, JP, NF, BP for biopharmaceutical production, SAFC®
Saltsyre
Formel: HCl
Molekylvægt: 36.46 g/mol
Smeltepunkt: –30 °C
Massefylde: 1.18 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
UN nummer: 1789
ADR: 8