Natriumhydroxid 5 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Caustic soda
1.37041.2500 1.37041.9025 1.37041.9950 1.37041.9200 1.37041.9958
1.37041.2500EA 3050 DKK
1.37041.2500 1.37041.9025 1.37041.9950 1.37041.9200 1.37041.9958
Natriumhydroxid 5 mol/l, EMPROVE® EXPERT for biopharmaceutical production, SAFC®
Sodium hydroxide
Formel: NaOH
Kogepunkt: 109 °C (1013 hPa)
Massefylde: ≤1,11 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
UN nummer: 1824
ADR: 8,