Saltsyre 25%, EMPROVE® EXPERT Ph. Helv. for biopharmaceutical production, SAFC®
Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Hydrochloric acid solution, Muriatic acid
1.37312.9025 1.37312.9950 1.37312.2500
1.37312.9025EA 0 DKK
1.37312.9025 1.37312.9950 1.37312.2500
Saltsyre 25%, EMPROVE® EXPERT Ph. Helv. for biopharmaceutical production, SAFC®
Saltsyre
Formel: HCl
Kogepunkt: 110 °C (1013 hPa)
Smeltepunkt: –30 °C
Massefylde: 1,18 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011324
CAS-nummer: 7647-01-0
EINECS: 231-595-7
UN nummer: 1789
ADR: 8,