Natriumhydroxid standardised solution 0.05 N in aqueous solution
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Caustic soda
35623.K2 35623.K7 35623.M4
35623.K2EA 447 DKK
35623.K2 35623.K7 35623.M4
Natriumhydroxid standardised solution 0.05 N in aqueous solution
Sodium hydroxide
CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Physical Form: Liquid Odour: Odourless MDL No.: MFCD00003548
Formel: NaOH
Molekylvægt: 40 g/mol
Kogepunkt: 1390
Smeltepunkt: 323
Massefylde: 1 g/cm³ (25 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00003548
CAS-nummer: 1310-73-2
EINECS: 215-185-5
UN nummer: 1824
ADR: 8 III
Merck Index: 14,08627