Eddikesyre 60%, EMPROVE® EXPERT for biopharmaceutical production
Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
1.37034.9025 1.37034.9950
1.37034.9025EA 6540 DKK
1.37034.9025 1.37034.9950
Eddikesyre 60%, EMPROVE® EXPERT for biopharmaceutical production
Eddikesyre
Formel: H₃CCOOH
Molekylvægt: 60.05 g/mol
Kogepunkt: 118
Smeltepunkt: 17
Flammepunkt: 38,5
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00036152
CAS-nummer: 64-19-7
EINECS: 200-580-7
UN nummer: 2790
ADR: 8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM