Eddikesyre 30%, EMPROVE® EXPERT Ph. Helv. for biopharmaceutical production, SAFC®
Leverandør: MERCK PRODUCTION CHEMICALS
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
Synonymer: Vinegar acid, Methanecarboxylic acid, Ethanoic acid
1.37047.9025 1.37047.2500 1.37047.9950 1.37047.9958
1.37047.9025EA 0 DKK
1.37047.9025 1.37047.2500 1.37047.9950 1.37047.9958
Eddikesyre 30%, EMPROVE® EXPERT Ph. Helv. for biopharmaceutical production, SAFC®
Eddikesyre
Formel: H₃CCOOH
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00036152
CAS-nummer: 64-19-7
EINECS: 200-580-7
UN nummer: 2790
ADR: 8,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ecdb21d8177dd197c843d4b5187410fee102541&vid=6c26d1716b942e60cb8b3ec0f91e6623fed5b3db&prid=web&clid=SAPDAM