(±)-3-Methyl-2-butanol ≥98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
DL-3-Methyl-2-butanol
L05347.14
L05347.22
L05347.14EA
365
DKK
L05347.14
L05347.22
(±)-3-Methyl-2-butanol ≥98%
(±)-3-Methyl-2-butanol
Purity: 98%
CAS No.: 598-75-4
Molecular Formula: C5H12O
Formula Weight: 88.15
Flash Point: 26°(78°F)
Refractive Index: 1.4090
Boiling Point: 112°
Beilstein Reference: 1718800
Merck Reference: 14,6031
UN No.: 1105
MDL No.: MFCD00004527
Formel:
C₅H₁₂O Kogepunkt: 112 °C Smeltepunkt: –117 °C Massefylde: 0,818 Flammepunkt: 26 °C (78 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004527 CAS-nummer: 598-75-4 EINECS: 209-950-2 UN nummer: 1105 ADR: 3,III Merck Index: 14,6031 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...