(±)-3-Methyl-1-pentanol 99+%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

335320050 335320250 335321000
ACRO335320050EA 292 DKK
ACRO335320050 ACRO335320250 ACRO335321000
(±)-3-Methyl-1-pentanol 99+%
(±)-3-Methyl-1-pentanol
Formel: C₆H₁₄O
Kogepunkt: 150…154 °C
Smeltepunkt: –70 °C
Massefylde: 0,823 g/cm³
Flammepunkt: 58 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00002937
CAS-nummer: 589-35-5
EINECS: 209-644-9
UN nummer: 1987
ADR: 3,III

Bestil nu

Specifikationer Testresultater

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4190 to 1.4210 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR