(±)-2-Methyl-3-pentanol ≥99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
L08015.09
L08015.18
L08015.09EA
1100
DKK
L08015.09
L08015.18
(±)-2-Methyl-3-pentanol ≥99%
(±)-2-Methyl-3-pentanol
Purity: 99%
CAS No.: 565-67-3
Molecular Formula: C6H14O
Formula Weight: 102.18
Flash Point: 46°(114°F)
Refractive Index: 1.4170
Boiling Point: 127-129°
Beilstein Reference: 1718952
UN No.: 2282
MDL No.: MFCD00004567
Formel:
C₆H₁₄O Kogepunkt: 127…129 °C Smeltepunkt: –53 °C Massefylde: 0,830 Flammepunkt: 46 °C (114 °F) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004567 CAS-nummer: 565-67-3 EINECS: 209-286-3 UN nummer: 2282 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...