DL-Cystein 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Amino-3-mercaptopropanoic acid, H-DL-Cys-OH, (±)-Cysteine

230120100 230120500
ACRO230120100EA 578 DKK
ACRO230120100 ACRO230120500
DL-Cystein 97%
DL-Cystein
Formel: C₃H₇NO₂S
Kogepunkt: 254 °C (1013 hPa)
Smeltepunkt: 225 °C (dec.)
Massefylde: 1,441 g/cm³ (25 °C)
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00004881
CAS-nummer: 3374-22-9
EINECS: 222-160-2
Merck Index: 12,02850

Bestil nu

Specifikationer Testresultater

Appearance White to off-white Powder
Infrared spectrum Conforms
Elemental Analysis 11.12 to 12.02 % N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR