Chloriodmethan ≥98% stabilised
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Stabilization:
Stabilised with copper
A11032.09
A11032.18
A11032.30
A11032.36
A11032.09EA
380
DKK
A11032.09
A11032.18
A11032.30
A11032.36
Chloriodmethan ≥98% stabilised
Chloroiodomethane
Stabilized with copper
Purity: 98%
CAS No.: 593-71-5
Molecular Formula: ClCH2I
Formula Weight: 176.38
Flash Point: None
Refractive Index: 1.5822
Boiling Point: 108-109°
Beilstein Reference: 1730802
UN No.: 2810
MDL No.: MFCD00001078
Formel:
CH₂ClI Kogepunkt: 108…109 °C Massefylde: 2,416 Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00001078 CAS-nummer: 593-71-5 EINECS: 209-804-8 UN nummer: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...