Chloracetone 96% Stabilised

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: Chloropropanone

173470010 173470250 173472500 173470050
173470010.EA 1170 DKK
173470010. 173470250. 173472500. 173470050.
Chloracetone 96% Stabilised
Chloracetone
Formel: C₃H₅ClO
Molekylvægt: 92.52 g/mol
Kogepunkt: 120
Smeltepunkt: –44.5 °C
Massefylde: 1.16 g/cm³
Flammepunkt: 32 °C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00000936
CAS-nummer: 78-95-5
EINECS: 201-161-1
UN nummer: 1695
ADR: 6.1 I
Merck Index: 13,02131

Bestil nu

Specifikationer Testresultater

Appearance Clear yellow to yellow-brown Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4330 (20°C, 589 nm)
GC ≥95 %
Stabilizer 0.1 % epoxidized soybean oil


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR