Polyethylen glycol 4.000

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: PEG 4000

A16151.0B A16151.0I A16151.30
A16151.0BEA 1040 DKK
A16151.0B A16151.0I A16151.30
Polyethylen glycol 4.000
Polyethylen glycol 4.000

CAS No.: 25322-68-3 Molecular Formula: H(OCH2CH2)nOH Odour: Odourless pH: 5-7 Melting Point: 58-62° Merck Reference: 14,7568 MDL No.: MFCD01779612

Formel: H(OCH₂CH₂)nOH
Molekylvægt: 4000 g/mol
Kogepunkt: ∼250 °C (1013 hPa)
Smeltepunkt: 58…62 °C
Massefylde: 1.13 g/cm³ (20 °C)
Flammepunkt: 250
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD01779612
CAS-nummer: 25322-68-3
EINECS: 500-038-2
Merck Index: 14,7568

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR