Sølv 1.000 mg/l i fortyndet salpetersyre (from AgNO3) ARISTAR® enkeltelement standard til ICP
Leverandør: VWR Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
456022KEA
684
DKK
456022K
456024M
Sølv 1.000 mg/l i fortyndet salpetersyre (from AgNO3) ARISTAR® enkeltelement standard til ICP
Standarder
Kationiske standarder
Silver Standards
AgNO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.