Bly 10.000 mg/l i fortyndet salpetersyre (from Pb(NO3)2) ARISTAR® enkeltelement standard til ICP
Leverandør: VWR Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
455572YEA
2220
DKK
455572Y
455574K
Bly 10.000 mg/l i fortyndet salpetersyre (from Pb(NO3)2) ARISTAR® enkeltelement standard til ICP
Standarder
Kationiske standarder
Lead Standards
Pb(NO₃)₂ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.