Ølgær

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

368080010 368080050 368080250
ACRO368080010EA 337 DKK
ACRO368080010 ACRO368080050 ACRO368080250
Ølgær
Ølgær
CAS-nummer: 68876-77-7

Bestil nu

Specifikationer Testresultater

Appearance Cream to brown Powder
Loss on drying ≤9 %
Protein 40 to 50 % (Typical value)
Microbial contamination (Salmonella) Absent in 375g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR