Platinum iridium alloy (90:10 w%) ≥99.9% (metals basis excluding precious metals), wire, annealed, Ø 0.127 mm (0.005 in)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
43019.BS
43019.BW
43019.BSEA
1030
DKK
43019.BS
43019.BW
Platinum iridium alloy (90:10 w%) ≥99.9% (metals basis excluding precious metals), wire, annealed, Ø 0.127 mm (0.005 in)
Platinum iridium alloy (90:10 w%)
Purity: 99.9%
Molecular Formula: Pt:Ir; 90:10 wt%
Physical Form: ≈0.03g/10cm, Temper: soft
Odour: Odourless
MDL No.: MFCD02091683
Vacuum evaporation deposition, scanning and tunneling microscopy, corrosion resistant electrodes