Platinum iridium alloy (90:10 w%) ≥99.9% (metals basis), foil, Thickness: 0.025 mm (0.001 in)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
41802.FF
41802.FI
41802.GH
41802.FFEA
3930
DKK
41802.FF
41802.FI
41802.GH
Platinum iridium alloy (90:10 w%) ≥99.9% (metals basis), foil, Thickness: 0.025 mm (0.001 in)
Platinum iridium alloy (90:10 w%)
Purity: 99.9%
Molecular Formula: Pt:Ir; 90:10 wt%
Physical Form: ≈0.34g/25x25mm
Odour: Odourless
MDL No.: MFCD02091683