Platinum iridium alloy (90:10 w%), wire, hard, Ø 0.5 mm (0.02 in)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
10056.BS
10056.BW
10056.CB
10056.BSEA
1860
DKK
10056.BS
10056.BW
10056.CB
Platinum iridium alloy (90:10 w%), wire, hard, Ø 0.5 mm (0.02 in)
Platinum iridium alloy (90:10 w%)
Molecular Formula: Pt:Ir; 90:10 wt%
Physical Form: ≈0.44g/10cm, Temper: as drawn
Odour: Odourless
MDL No.: MFCD02091683