Platinum iridium alloy (80:20 w%), wire, hard, Ø 1.0 mm (0.04 in)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
44732.BQ
44732.BS
44732.BW
44732.BQEA
7750
DKK
44732.BQ
44732.BS
44732.BW
Platinum iridium alloy (80:20 w%), wire, hard, Ø 1.0 mm (0.04 in)
Platinum iridium alloy (80:20 w%)
Molecular Formula: Pt:Ir; 80:20 wt%
Physical Form: ≈0.85g/5cm
Odour: Odourless
MDL No.: MFCD02091683