Platinum iridium alloy (80:20 w%) ≥99.9% (metals basis), foil, Thickness: 0.15 mm (0.006 in)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
43813.GH
43813.FI
43813.GHEA
0
DKK
43813.GH
43813.FI
Platinum iridium alloy (80:20 w%) ≥99.9% (metals basis), foil, Thickness: 0.15 mm (0.006 in)
Platinum iridium alloy (80:20 w%)
Purity: 99.9%
Molecular Formula: Pt:Ir; 80:20 wt%
Physical Form: ≈8.13g/50x50mm
Odour: Odourless
MDL No.: MFCD02091683