Platinum iridium alloy (80:20 w%) ≥99.9% (metals basis), tråd, annealed, Ø 0.2 mm (0.008 in)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
39526.BS
39526.BW
39526.BSEA
1720
DKK
39526.BS
39526.BW
Platinum iridium alloy (80:20 w%) ≥99.9% (metals basis), tråd, annealed, Ø 0.2 mm (0.008 in)
Platinum iridium alloy (80:20 w%)
Purity: 99.9+%
Molecular Formula: Pt:Ir; 80:20 wt%
Physical Form: ≈0.7g/10cm, Temper: soft
Odour: Odourless
MDL No.: MFCD02091683