Palladium silver alloy (75:25 w%) ≥99.9% (metals basis excluding Pt), foil, Thickness: 0.025 mm (0.001 in)
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
42187.FI
42187.GH
42187.FIEA
14260
DKK
42187.FI
42187.GH
Palladium silver alloy (75:25 w%) ≥99.9% (metals basis excluding Pt), foil, Thickness: 0.025 mm (0.001 in)
Palladium silver alloy (75:25 w%)
Purity: 99.9%
Molecular Formula: Pd:Ag; 75:25 wt%
Physical Form: ≈0.18g/25x25mm
Odour: Odourless
MDL No.: MFCD02091738