Acrylamid-Bisacrylamid (19:1) 40% in aqueous solution
Leverandør: Serva
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
Synonymer:
Acrylamid-Bis
10679.01
10679.02
10679.03
SERA10679.01EA
313.26
DKK
SERA10679.01
SERA10679.02
SERA10679.03
Acrylamid-Bisacrylamid (19:1) 40% in aqueous solution
Acrylamid-Bis (32:1)
Massefylde:
1.03 g/cm³ (20 °C) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00080848 UN nummer: 3426 ADR: 6.1 III |