Stivelse, opløselig ACS for iodometric titrations
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Starch, Vitex® iodine indicator, Amylodextrin
36703.18
36703.30
36703.A1
36703.18EA
481
DKK
36703.18
36703.30
36703.A1
Stivelse, opløselig ACS for iodometric titrations
Stivelse, opløselig
CAS No.: 9005-84-9
Physical Form: Powder
Odour: Odourless
Melting Point: 256-258° dec.
Solubility: Soluble in hot water
Merck Reference: 14,8799
MDL No.: MFCD00082026
Formel:
(C₆H₁₀O₅)n Smeltepunkt: 256…258 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00082026 CAS-nummer: 9005-84-9 EINECS: 232-686-4 Merck Index: 14,8799 |