Cellulose

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

14204.02 14204.03
SERA14204.02EA 507.29 DKK
SERA14204.02 SERA14204.03
Cellulose
Cellulose
Formel: (C₁₂H₂₂O₁₁)n
Smeltepunkt: 260…270 °C
Massefylde: 1,27…1.6 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00081512
CAS-nummer: 9004-34-6
EINECS: 232-674-9
Merck Index: 13,01977

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR