Gentamicin sulfat

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: NMD Inhibitor II, Garamycin, Gentamicin sulphate salt, Gentiomycin C

22185.01 22185.02 22185.03
SERA22185.01EA 126.8 DKK
SERA22185.01 SERA22185.02 SERA22185.03
Gentamicin sulfat
Gentamicin sulfat
Smeltepunkt: 218…237 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00270181
CAS-nummer: 1405-41-0
EINECS: 215-778-9
Merck Index: 13,04403

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR