D(+)-Sucrose Reag. Ph. Eur., Millipore®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, Sucrose
M-Clarity™ quality level = MQ100
1.07687.0250 1.07687.1000 1.07687.5000
1.07687.0250EA 253 DKK
1.07687.0250 1.07687.1000 1.07687.5000
D(+)-Sucrose Reag. Ph. Eur., Millipore®
D(+)-Sucrose
Formel: C₁₂H₂₂O₁₁
Molekylvægt: 342.3 g/mol
Smeltepunkt: 169…170 °C
Massefylde: 1.29 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00006626
CAS-nummer: 57-50-1
EINECS: 200-334-9
Merck Index: 13,08966
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=971b1a9d1f02c06efd63ac665dfb191799fbd5ac&vid=2ddc1affd401bacf02244ae0cb620ff18e440965&prid=web&clid=SAPDAM