D(+)-Trehalose dihydrat

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Mycose, α,α-Trehalose, 1-O-α-D-Glucopyranosyl-α-D-glucopyranoside, α-D-Glucopyranosyl-α-D-glucopyranoside, (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-Oxybis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) dihydrate

36770.02 36770.03
SERA36770.02EA 328.42 DKK
SERA36770.02 SERA36770.03
D(+)-Trehalose dihydrat
D(+)-Trehalose dihydrat
Formel: C₁₂H₂₂O₁₁·2H₂O
Smeltepunkt: 97…99 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00071594
CAS-nummer: 6138-23-4
EINECS: 202-739-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af741ee934bfe7c82395d31c21e2c19f0738d0f0&vid=59183c854dcfa367e15d46861ef4b9a7028540a9&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR