Fluoro-Jade C (FJC), RTD™ Ready-to-Dilute Staining Kit for identifying Degenerating Neurons
Leverandør: Biosensis
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
TR-100-FJ TR-100-FJT
BSENTR-100-FJEA 4730 DKK
BSENTR-100-FJ BSENTR-100-FJT
Fluoro-Jade C (FJC), RTD™ Ready-to-Dilute Staining Kit for identifying Degenerating Neurons
Fluoro-Jade
Fluoro-Jade stain is a fluorochrome derived from fluorescein, and is commonly used in neuroscience disciplines to label degenerating neurons in ex vivo tissue of the central nervous system.
The causes and effects of neuronal degeneration are of major interest to a wide variety of neuroscientists. Paralleling this growing interest is an increasing number of methods applicable to the detection of neuronal degeneration. Fluoro-Jade® C stains all degenerating neurons regardless of specific insult or mechanism of cell death. Fluoro-Jade C exhibits the greatest signal to background ratio, as well as the highest resolution. This translates to a stain of maximal contrast and affinity for degenerating neurons. This makes it ideal for localising not only degenerating nerve cell bodies but also distal dendrites, axons and terminals. The dye is highly resistant to fading and is compatible with virtually all histological processing and staining protocols. Reagent detects degenerating neurons, and neuronal degeneration. There is no specific staining in normal healthy brain. Note: Some researchers under some conditions report blood vessel staining with Fluoro Jade. This may be because Fluoro Jade is an analogue of eosin (which stains blood cells). In general, good perfusion and preparation of the tissue should help prevent blood vessel staining but it may not be possible to eliminate it entirely. In our experience it is generally possible to distinguish neuronal from blood vessels staining by eye.
The Fluoro-Jade C 'Ready to Dilute' (RTD™) Staining Kit provides an easy to use assortment of Fluoro-Jade C, DAPI, sodium hydroxide and potassium permanganate in liquid form. Following our detailed protocol, Fluoro-Jade C labelled degenerating neurons are visualised with blue light excitation while DAPI counter stained cell nuclei are visualised with ultra-violet illumination. The Fluoro-Jade C Staining Kit can be used on all kinds of preserved tissues, including fresh-frozen, paraformaldehyde or formalin fixed, and formalin fixed, paraffin-embedded tissues. FJC visualization is accomplished using blue light or a 488nm Laser. Excitation Peak: 495 nm Emission Peak: 521 nm Filter system for visualizing: Fluorescein/FITC
MATERIALS PROVIDED
Sodium Hydroxide, Solution A (Dilute 1:10 prior to use) - 40 mL
Potassium Permanganate, Solution B (Dilute 1:10 prior to use) - 40 mL
Fluoro-Jade C, Solution C (Dilute 1:10 prior to use) - 40 mL
DAPI, Solution D (Add to diluted Fluoro-Jade C) - 40 mL