Phenanthren-D10 (98% D)
Leverandør: CAMBRIDGE ISOTOPE LABS
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Phenanthrene-[²H₁₀]
DLM-371-0.1
DLM-371-1
CAMBDLM-371-0.1EA
0
DKK
CAMBDLM-371-0.1
CAMBDLM-371-1
Phenanthren-D10 (98% D)
Phenanthren-D10
Formel:
C₁₄D₁₀ |
CAS-nummer:
1517-22-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...