Chemical process indicators
Leverandør: CROSSTEX
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
CPI-E01 CPI-E02 SP-E16
NAMSCPI-E01EA 1910 DKK
NAMSCPI-E01 NAMSCPI-E02 NAMSSP-E16
Chemical process indicators
Sterilisationsindikatorer
Chemical process indicators provide a fast and consistent visual check on items that were sterilised by EO.
  • Consistent and irreversible colour transition
  • Permanent pressure-sensitive adhesive
  • Indicator inks do not contain lead or other toxic heavy metals
Chemical indicator: Initial colour (purple) and signal colour (green)
Storage condition: 15 to 30 °C
Relative humidity: 20 – 70%
Certificeringer: Compliance to ISO 11140-1:2014.