Dichlormethan-D2 (99.9% D)

Leverandør: CAMBRIDGE ISOTOPE LABS
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Dichloromethane-d2, Methylene chloride-D2, Dichloromethane-[²H₂], Dideuteromethylene chloride, Methylene chloride-d2

DLM-23-10 DLM-23-10X0.75 DLM-23
CAMBDLM-23-10EA 0 DKK
CAMBDLM-23-10 CAMBDLM-23-10X0.75 CAMBDLM-23
Dichlormethan-D2 (99.9% D)
Dichlormethan-D2
Formel: CD₂Cl₂
Molekylvægt: 86.92 g/mol
Kogepunkt: 40
Smeltepunkt: –97 °C
Massefylde: 1.36 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00000882
CAS-nummer: 1665-00-5
EINECS: 216-776-0
UN nummer: 1593
ADR: 6.1 III

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR