Petroleumbenzin, 80…110 °C

Leverandør: Roth Carl
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

3259.1 3259.2
ROTH3259.1EA 0 DKK
ROTH3259.1 ROTH3259.2
Petroleumbenzin, 80…110 °C
Petroleumbenzin
Kogepunkt: 80…110 °C (1013 hPa)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00081849
CAS-nummer: 64742-49-0
EINECS: 265-151-9
UN nummer: 1268
ADR: 3,II

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR