Paraffin, Solidification point: 46-48°C, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer: Wax Paraffin, Paraffin wax
M-Clarity™ quality level = MQ200
1.07151.1000 1.07151.9025
1.07151.1000EA 923 DKK
1.07151.1000 1.07151.9025
Paraffin, Solidification point: 46-48°C, Supelco®
Paraffin
Massefylde: 0.9 g/cm³ (20 °C)
CAS-nummer: 8002-74-2
EINECS: 232-315-6