Natronkalk, pellets analytical reagent, with indicator
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Farlig
M-Clarity™ quality level = MQ200
1.06839.5001 1.06839.9025 1.06839.1001
1.06839.5001EA 4800 DKK
1.06839.5001 1.06839.9025 1.06839.1001
Natronkalk, pellets analytical reagent, with indicator
Soda lime
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00134094
CAS-nummer: 8006-28-8
UN nummer: 1907
Merck Index: 13,08640
Bestil nu
Specifikationer Testresultater
Identity passes test
Absorption capacity for CO₂ (method NF XVII) ≥ 25 %
Loss on Drying (105°C) 13.0 - 18.0 %
Color change passes test
Bulk density about 75
Particle size (2 - 5 mm) ≥ 95 %