Nikkel ≥99.98% (metals basis), wire, annealed, Ø 2.0 mm (0.08 in)

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

43134.G6 43134.G2
43134.G6EA 5820 DKK
43134.G6 43134.G2
Nikkel ≥99.98% (metals basis), wire, annealed, Ø 2.0 mm (0.08 in)
Nikkel

Purity: 99.98% CAS No.: 7440-02-0 Physical Form: ≈28.0g/m, Temper: soft Odour: Odourless MDL No.: MFCD00011137

Formel: Ni
Kogepunkt: 3007 °C (1013 hPa)
Smeltepunkt: 1453 °C
Massefylde: 8,9 g/cm³ (20 °C)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00011137
CAS-nummer: 7440-02-0
EINECS: 231-111-4
UN nummer: 3089
ADR: 4.1,III
Merck Index: 12,06582

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR